Would you like to log in again?
American Chemical Society: Glow It Up Lab Activity
Supplier: ALDON CORP SE
IS2552
470310-420KT 74.9 USD InStock
470310-420
American Chemical Society: Glow It Up Lab Activity
Educational Materials Chemistry Educational Materials Electrochemistry Learning Activities
How does a glow stick work?
  • Chemiluminescence
  • Students alter the chemical reaction to attempt to change duration and brightness of the reaction
This lab, developed by the American Chemical Society through grants from the National Science Foundation (NSF) and National Institutes of Health (NIH), explores chemical reactions that release energy in the form of light, known as chemiluminescence. Teachers begin with a demonstration that uses a luminol mixture and oxidizer to start a chemical reaction that glows.
Students will then conduct an experiment to see how the amount of oxidizer affects the brightness and duration of the light. Students will also add a fluorescent highlighter dye to the reaction to see how this affects the brightness and duration of the glow. Additionally, they will learn about the invention of the glow stick and relate their experience to how a glow stick works.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty