Would you like to log in again?
American Chemical Society: CO2 to The Rescue Lab Activity
Supplier: ALDON CORP SE
IS2553
470310-422KT 110.3 USD InStock
470310-422
American Chemical Society: CO2 to The Rescue Lab Activity
Educational Materials Chemistry Educational Materials Gas Analysis Learning Activities
Invent a small device to rescue a cell phone from water.
  • Chemical reaction
  • Stoichiometry
  • Design challenge
This series of activities, developed by the American Chemical Society through grants from the National Science Foundation (NSF) and National Institutes of Health (NIH), is comprised of a lesson that begins with a design challenge: to invent a small device that could rescue a cell phone that accidentally falls into water. The teacher starts off by showing students a balloon that inflates with carbon dioxide gas as chemicals inside the balloon react with one another.
Students conduct a pair of chemical reactions to determine which of two acids react with baking soda to produce the most carbon dioxide gas. Once students determine the best acid to use, they compare the amount of gas produced with different amounts of baking soda. Finally, trying to use the smallest volumes possible, students discover how much of each reactant is needed to fully inflate a small zip-closing plastic bag to see if it can get a model clay cell phone to float.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty