Would you like to log in again?
Phone Scope Magnifier, 60×
Supplier: L&W OPTICS ELECTRONICS LTD SE
LVA031
470310-434CS 2240.8 USD InStock
470310-434
Phone Scope Magnifier, 60×
Magnifiers
Quick and easy to attach to a phone's camera with the detachable clip.
  • 60× Zoom microscope magnifing lens for universal phones
  • Bright LED light of the magnifying glass provides enough light at any dim condition
  • On/Off switch for LEDs
  • Portable and convenient design
  • Aligned with GLOBE protocols
This fits many types of cell phones and tablets. Look at images on a cell phone’s large screen, zoom in and take pictures to share with others or input into documents or other media. Quickly attaches to a phone's camera port with the detachable clip.
Perfect for biological, jewelry, coins, currency, stamps, antiques, precision parts, insect and flower photography, geology, horticulture plant identification take up-close pictures of plant issues and send them to others for identification.
Delivery information: LEDs powered by 3 LR1130 cell batteries are included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty