Would you like to log in again?
Crazy Circuits Classroom Sets: Programming 101
Supplier: BROWN DOG GADGETS
Crazy Circuits Ward's®
K1051-01PK K1051-10PK K1051-25PK K1051-05PK K1032-24PK K1032-12PK
470311-068EA 2263.9 USD InStock
470311-068 470348-914 470348-918 470311-070 470348-922 470348-942
Crazy Circuits Classroom Sets: Programming 101
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A complete classroom solution to the daunting task of teaching electronics and programming.
  • As with all Crazy Circuits kits, our classroom sets require no soldering
  • Parts are connected in a brick-based building environment via our special maker tape conductive tape
  • The only additional tools you'll need are scissors and a computer to manipulate code
The Programming 101 Classroom line was designed as a complete classroom solution for teaching electronics. These modular kits are robust sets of materials supported by online lesson plans, projects, activities, diagrams, and videos. Programming 101 sets each come with one Teacher box and your choice of 4, 9, or 24 Student boxes filled with electronics parts and supplies. The Teacher box includes a full set of Student supplies as well as additional materials. These modular kits are supported by equally robust, NGSS-aligned, online teaching resources designed to facilitate easy classroom use. In addition to lesson plans, the online resources also include activity pages, assessments, diagrams, and videos for classroom use.
Ordering information: Scissors and computer(s) are required but not included.
Delivery information: Each Student box includes 42 Parts + organizer, (5) Switches (pushbutton, slide), (4) LEDs, (4) Resistors, (4) Brick plates, (3) Ribbon wire sets, (2) Female headers, (2) Potentiometers, (1) Roll ⅛” Maker Tape™, (1) Photoresistor, (1) Neopixel, (1) Neopixel strip, (1) Programmable robotics (Arduino™) board, (1) LEGO™ compatible servo + connector, (1) Ultrasonic distance sensor, (1) 4-Bit 7 segment display, (1) Joystick, (1) Temperature/humidity sensor, (1) Brick baseplate, (1) USB cable, (1) Bundle of 5 alligator clips, Instructions cards/online curriculum. Teacher box includes (1) Complete contents of Student box, (1) Large organizer (replaces organizer from Student box), (4) additional rolls of ⅛" Nylon conductive tape, (2) Additional female to female ribbon wire sets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty