Would you like to log in again?
Quantitative Determination of Carbon, Leybold Products
Supplier: Leybold
C2.1.1.1
470311-202EA 5765.99 USD InStock
470311-202
Quantitative Determination of Carbon, Leybold Products
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Leybold Chemistry Experiments.
  • Determine the composition of organic compounds
  • Learn about quantitative analysis
  • Analyze the proportion of carbon in a compound
  • Determine the structure of organic compounds
  • Learn an example of the application of a redox reaction
In experiment C2.1.1.1, the carbon content of an organic compound is determined quantitatively. In a combustion analysis, the substance is passed over glowing copper(II) oxide at approx. 800 to 900 °C. The carbon is converted to CO2 and the hydrogen is converted to H2O. The CO2 gas generated is collected in a gas syringe and the volume is measured.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty