Would you like to log in again?
Fractionated Petroleum Distillation, Leybold Products
Supplier: Leybold
C2.3.2.1
470311-208EA 10604.09 USD InStock
470311-208
Fractionated Petroleum Distillation, Leybold Products
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Leybold Chemistry Experiments.
  • Work with and investigate crude oil
  • Preform a fractionated distillation of crude oil
  • Learn about bubble tray column
  • Relate the structure to the properties of chemical compounds
Crude oil is first separated into different fractions by fractionated distillation. Individual substances cannot be isolated in this way, because the boiling points are very close to one another. Rather, the objective here is to collect hydrocarbons of certain boiling point ranges.
In chemical engineering, crude oil is distilled in bubble tray columns. This process is simulated in experiment C2.3.2.1. The system is fitted with two bubble trays so multiple fractions can be extracted simultaneously.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty