Would you like to log in again?
Recording of Absorption Spectra, Leybold Products
Supplier: KLINGER EDUCATION PROD CORP SE
C3.3.1.2
470311-210EA 9768.69 USD InStock
470311-210
Recording of Absorption Spectra, Leybold Products
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Leybold Chemistry Experiments.
  • Observe and understand light spectra
  • Recognize connection between absorption, reflection and transmission
  • Appreciate color as light of characteristic wavelengths
  • Identify the interaction of light with colored liquids
In experiment C3.3.1.2, the solution of a dye is exposed to the light from a lamp. For the absorption spectrum, the light passing through the colored solution is recorded with a spectrometer. The absorption spectrum is compared with the continuous spectrum of the light from the lamp.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty