Would you like to log in again?
Reaction of Malachite Green, Leybold Products
Supplier: Leybold
C4.1.3.1
470311-212EA 7697.79 USD InStock
470311-212
Reaction of Malachite Green, Leybold Products
Educational Materials Chemistry Educational Materials Thermochemistry/Kinetics Learning Activities
Leybold Chemistry Experiments.
  • Reaction kinetics
  • Influencing the rate of reaction
  • Chemical equilibrium
  • Transport processes
Reactions in which a participant in the reaction changes color are a suitable choice if one wishes to observe the course of a reaction. Such reactions are easy to track photometrically, because the decrease and increase in the extinction rate can be directly converted mathematically into concentration changes.
In experiment C4.1.3.1, the reaction of malachite green with hydroxide ions is monitored with an immersion photometer in a glass beaker. Blue-green at first, the solution loses color when sodium hydroxide is added. This simple system makes it possible to investigate how the rate of reaction is affected by temperature, concentration and the choice of solvent.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty