Would you like to log in again?
Leybold Determination of the Molar Mass of Gases
Supplier: Leybold
C1.1.1.2
470311-216EA 9519.59 USD InStock
470311-216
Leybold Determination of the Molar Mass of Gases
Educational Materials Chemistry Educational Materials Gas Analysis Learning Activities
Leybold Chemistry Experiments.
  • Show that gases also have a weight
  • Determine the molecular mass of gases
  • Learn about the molar volume
At constant pressure and constant temperature, any gas occupies the same volume regardless of the type of atom or the composition. So if we know the volume, pressure and temperature, we can then determine the molar mass of gases. The measurement is conducted in experiment C1.1.1.2 with a glass sphere for weighing gases.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty