Would you like to log in again?
Melting Ice, Boiling Water, Leybold Products
Supplier: Leybold
C1.1.2.1
470311-218EA 4156.29 USD InStock
470311-218
Melting Ice, Boiling Water, Leybold Products
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
Leybold Chemistry Experiments.
  • Understanding the various states of matter
  • Transition of substances between different aggregate states
  • Melting of ice (transition solid - liquid)
  • Boiling of water (transition liquid - gaseous)
Experiment C1.1.2.1 examines the behavior of water at different temperatures. To that end, ice (solid water) is slowly heated until it melts and then evaporates. At the boiling point and the melting point, the temperature does not change until the substance has transitioned completely to the other state. The boiling point and melting point are easy to determine in this way.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty