Would you like to log in again?
Estimation of the Size of Oil Molecules, Leybold Products
Supplier: Leybold
C1.1.3.2
470311-220EA 752.49 USD InStock
470311-220
Estimation of the Size of Oil Molecules, Leybold Products
Educational Materials Chemistry Educational Materials Atomic Structure Learning Activities
Leybold Chemistry Experiments.
  • Determination of molar mass
  • States of matter
  • Analysis of Rutherford scattering
  • Chemical reactions and stoichiometry
One of the important questions in atomic physics is the question of the size of the atom. The study of the size of molecules provides experimentally easier access on a manageable magnitude. Experiment C1.1.3.2 uses simple means to estimate the size of molecules from the size of a patch of oil floating on the surface of water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty