Would you like to log in again?
LEYBOLD® Hydrogen As A Reducing Agent
Supplier: Leybold
C1.4.3.1
470311-230EA 6231.09 USD InStock
470311-230
LEYBOLD® Hydrogen As A Reducing Agent
Educational Materials Chemistry Educational Materials Gas Analysis Learning Activities
LEYBOLD® Chemistry Experiments.
  • Chemical reactions and stochiometry
  • Components of air
  • Production of gases
  • Gas laws
Copper oxide is reduced with hydrogen to form elementary copper. Hydrogen is a good reducing agent. Its ability to react applies not only to free oxygen but also to oxygen compounds. The metal is formed in this way when transforming a metal oxide (as in this experiment) with hydrogen.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty