Would you like to log in again?
LEYBOLD® Temperature-Dependency of the Pressure of Gas at A Constant Volume
Supplier: Leybold
C1.4.4.3
470311-236EA 5948.59 USD InStock
470311-236
LEYBOLD® Temperature-Dependency of the Pressure of Gas at A Constant Volume
Educational Materials Chemistry Educational Materials Thermochemistry/Kinetics Learning Activities
LEYBOLD® Chemistry Experiments.
  • Kinetic theory of gases
  • Gas laws
  • Heat as a form of energy
  • Specific heat of gases
The pressure P in the air column is constantly reduced by evacuating the air at the open end so that the volume V of the air column also remains constant as the temperature drops. This experiment measures the pressure p of the air column as a function of the temperature T of the water bath. The evaluation confirms the relationship.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty