Would you like to log in again?
STEM Investigations: Fingerprint Classification
Supplier: ALDON CORP SE
IS3451
470311-678KT 99.25 USD InStock
470311-678
STEM Investigations: Fingerprint Classification
Educational Materials Forensics Educational Materials Fingerprint Analysis Learning Activities
Real world STEM investigation.
  • Additional optional activities included
  • Students create classification system
In this lab activity students will learn about the history of fingerprinting as a tool for identification. They then will analyze fingerprints for common characteristics and create a classification system using those characteristics.
Once they have created a system, they will analyze class data using the system that is used worldwide for classification. This kit includes an optional activity where students can practice lifting a fingerprint and comparing it to a 'crime scene' print. Kit contains enough materials for 15 groups. Teacher’s guide and student guide copymasters are included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty