Would you like to log in again?
3B Scientific® Diabetic Foot Trainer
Supplier: AMERICAN 3B SCIENTIFIC SE
3B Scientific®
1018815
470311-926EA 227.1 USD InStock
470311-926
3B Scientific® Diabetic Foot Trainer
Models Human Biology Models
Identify problems caused by diabetes.
  • Shows gangrenous toe, trophic ulcer, and pressure ulcer
  • Latex free
  • Three years manufacturers warranty
The after-effects of a diabetes mellitus can result in defects and pressure points in the area of the foot. In this Decubitus Foot skill trainer, a gangrenous alteration on the big toe of the right foot has already spread from the back of the foot to the metatarsal.
A mal perforans (trophic ulcer) is visible on the sole of the foot (plantar). A grade 3 pressure-ulcer is depicted in the heel area. A complete skin layer is lost and the subcutaneous tissue is damaged up to necrosis. This is recognizable by black coloration in the wound area.
Delivery information: Leg not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty