Would you like to log in again?
3B Scientific® Imaging Guided Lumbar Injection Trainer
Supplier: AMERICAN 3B SCIENTIFIC SE
3B Scientific®
1021898
470311-930EA 6760.29 USD InStock
470311-930
3B Scientific® Imaging Guided Lumbar Injection Trainer
Models Human Biology Models
Use for live imaging guidance.
  • Life-like radiopacity for realistic x-ray images
  • Realistic injection haptics
  • Anatomically accurate bone structure
  • Visually identifiable landmarks
The Image Guided Lumbar Spinal Injection Trainer enables trainees to develop a three-dimensional understanding of the procedures for successful spine interventions. They will learn to correlate imaging with the lumbar spinal anatomy relevant to interventional pain procedures, to identify the target tissue and vulnerable structures using imaging and anatomic inspection, and to apply best practices to ensure patient safety during the interventional spine procedures. Using the Spinal Injection Trainer is an economic alternative for imaging techniques courses on cadavers and offers the instructors a reliable, standardized patient simulation always ready to use.
The following image guided spinal interventions can be trained on the simulator:
Transforaminal Epidural Steroid Injections (TFSI)
Intralaminar Epidural Injection
Facet Blocks
Medial Branch Block (MBB/RF)
S1-block
Sacroiliac Joint Injection (SIJ-injection)
The Lumbar Spinal Injection Trainer depicts the following bone structures:
Sacrum, S1-S3 (with sacral hiatus canal)
Ilium, bilateral without ischial bone and hip joint
T12-L5
Coccyx
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty