Would you like to log in again?
3B Scientific® Physical Eye Model
Supplier: AMERICAN 3B SCIENTIFIC SE
3B Scientific®
1003806
470311-948EA 2051.65 USD InStock
470311-948
3B Scientific® Physical Eye Model
Models Zoology Models
How does an eye work?
  • Examine eye function and the interaction of lenses
  • Identify common problems like near and far sightedness
  • Includes case for storage or transport
This physical eye model can be used to demonstrate the optical functions of the human eye, e.g. representation of an object on the retina, accommodation (change in the lens curvature), short-sightedness and far-sightedness.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty