Would you like to log in again?
Mammal Bone Comparisons
Supplier: AMERICAN 3B SCIENTIFIC SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1021042 1021044 1021043
470312-594EA 2182.2 USD InStock
470312-594 470312-596 470312-598
Mammal Bone Comparisons
Models Zoology Models
High quality natural bone.
Hind limbs: Included are four real bone specimens: the hind leg of a dog and the hind foot of a horse, a cow and a pig respectively, as well as an original plastic cast of a human foot.
Fore limbs: Included are four real bone specimens: the front leg of a dog and the front foot of a horse, a cow and a pig respectively, as well as an original plastic cast of a human hand.
Teeth: A comparison of the different tooth shapes and types allows conclusions to be drawn about the various sources of nutrition as well as the feeding and hunting behavior of omnivores (pigs), carnivores (cats, dogs) and herbivores (cows, rabbits, rats).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty