Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Mammal Bone Comparison
Supplier: AMERICAN 3B SCIENTIFIC SE
1021068 1021066 1021065 1021067 1021047
470312-630EA 399.6 USD InStock
470312-630 470312-622 470312-624 470312-626 470312-628
Mammal Bone Comparison
Models Zoology Models
Comparison of mammal metatarsal bones: Includes four longitudinal sections of the following mammal metatarsal bones: horse, cow, pig, and chicken. The longitudinal section makes it possible to clearly explain the stability of the bones and to allow conclusions about the animal's locomotion.
  • High quality natural bone
Femur: Real femur of a mammal to demonstrate the typical bone structure and anatomy.
Humerus: Real humerus of a mammal to demonstrate the typical bone structure and anatomy.
Metacarpal: Real metacarpal of a mammal to demonstrate the typical bone structure and anatomy.
Metatarsal: Real metatarsal of a mammal to demonstrate the typical bone structure and anatomy.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty