Would you like to log in again?
Ward's® University Rock Collection
Supplier: Avantor
Ward's®
WARD470313-078 WARD470313-108
470313-108KT 6143 USD InStock
470313-108 470313-106 470313-078
Ward's® University Rock Collection
Educational Materials Earth Science Educational Materials Minerals and Rocks Learning Activities
Our most comprehensive collection of rock samples.
One of a kind collection
100 samples of igneous, sedimentary, and metamorphic rocks from around the world
Each sample is numbered and includes a corresponding identification list
Hand samples are 3×4" and are housed in individual, labeled plastic trays. The thin sections are housed in a slide storage box and are an outstanding addition for college petrographic studies.
Ordering information: Order the hand specimen set, thin section set, or matching hand specimen and thin section set.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty