Would you like to log in again?
Glass Distillation Set, 15 Piece, Eisco Scientific
Supplier: EISCO SCIENTIFIC LLC
CH2019003
470313-166EA 687.1 USD InStock
470313-166
Glass Distillation Set, 15 Piece, Eisco Scientific
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Distillation kit includes 15 pieces of glassware made from high quality, heavy duty, 3.3 borosilicate glass.
  • Heavy duty, 3.3 borosilicate glass
  • All pieces have 19/22 tapered joints
  • Supplied in hard-shell case with a custom foam insert for convenient and safe storage
  • Case measures 19" long, 16" wide, a 5" deep; features a locking system, handle, and rubber stoppers
High quality glassware set designed for small scale organic chemistry distillation experiments and procedures. All of the glassware is made from high quality, 3.3 borosilicate glass and feature 19/22 tapered joints ensuring a perfect fit.
Delivery information: Set includes: 1 condensor, 1 distilling column, 1 glass bleed tube, 1 thermometer adaptor, (1) 3-way adaptor, 1 Claisen adaptor, 1 vacuum adaptor, (1) 125 ml separating funnel, (1) 500 ml round bottom flask, (1) 250 ml round bottom flask, (1) 100 ml round bottom flask, (1) 50 ml round bottom flask, (1) 25 ml round bottom flask, (1) glass stopper.
Packaging: The complete kit is packaged in a hard shell case that includes a custom foam insert. This ensures safe delivery and storage of your set. The case also features a handle, lock, and rubber feet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty