Would you like to log in again?
Ceramic Heat Pad
Supplier: GSC INTERNATIONAL SE
CP-12
470313-334EA 11.65 USD InStock
470313-334
Ceramic Heat Pad
Surface Protectors Bench Protectors
These heat-insulating pads with low thermal conductivity are made from flexible aluminum and silica oxides with a small amount of organic binder.
  • Working temperature: 1000 °C or 1832 °F
  • Max. temperature: 1260 °C or 2300 °F
  • Sold as Each or as Pack of 12
They are useful for protecting surfaces when working with hot items like crucibles or flasks.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty