Would you like to log in again?
Glass Petri Dishes
Supplier: GSC INTERNATIONAL SE
1500-4-10 1500-3-10
470313-348CS 270.75 USD InStock
470313-348 470313-346
Glass Petri Dishes
Culture Dishes Petri Dishes
These glass petri dishes are two-piece (lid and dish) and are packed individually.
  • Covered dishes are not airtight
  • Transparent and reusable
  • Resistant to most acids, bases and alcohols
  • Not autoclavable
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty