Would you like to log in again?
Porcelain Crucibles and Covers
Supplier: GSC INTERNATIONAL SE
CHF-50-20 CLF-50-20 CHF-50-LID-20 CHF-30-LID-20 CLF-15-LID-20 CHF-20-20 CLF-30-20 CLF-15-20 CHF-10-20 CHF-15-LID-20 CHF-15-20 CHF-30-20
470313-408PK 33 USD InStock
470313-408 470313-406 470313-404 470313-402 470313-454 470313-356 470313-400 470313-452 470313-354 470313-398 470313-450 470313-396
Porcelain Crucibles and Covers
Crucibles
These crucibles are great for student use.
  • High and low form available
Crucibles are uniform and dense. They have stable mechanical strength and little permeability, and they are available in both high form and low form. When properly used, the glazed, high form pieces withstand temperatures of 1050 °C. Unglazed, low form pieces will withstand temperatures of 1350 °C.
Ordering information: Replacement covers are also available.
Packaging: Sold in packs of 20.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty