Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Pipette Pumps
Supplier: GSC INTERNATIONAL SE
PIPFL-25-10 PIPFL-10-10 PIPFL-2-10
470313-438EA 24.95 USD InStock
470313-438 470313-436 470313-324
Pipette Pumps
Pipettes Pipette Bulbs
Ideal alternative to pipette bulbs.
  • Easy, one-handed operation
  • Multiple capacity options
  • Easy to insert and remove
Pipette pumps are essential for filling pipettes with the accuracy needed in the lab. These pumps grip pipettes tightly, have both a thumb-wheel for precision in delivery and a quick release option, and are easily cleaned. This pump can hold up to 2 ml of fluid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty