Would you like to log in again?
Graduated Measuring and Volumetric Pipettes, Polypropylene
Supplier: Dynalon
303005-0050 303005-0005 302765-0001 302765-0010 303005-0001 303005-0010 302765-0005 303005-0025 302765-0002 303005-0002
470313-788CS 279.15 USD InStock
470313-788 470313-888 470313-786 470313-896 470313-894 470313-784 470313-892 470313-782 470313-890 470313-790
Graduated Measuring and Volumetric Pipettes, Polypropylene
Pipettes Graduated Pipettes
Disposable pipettes perfect for every lab use.
  • Available in graduated or volumetric styles
  • Available in variety of sizes
Translucent polypropylene graduated measuring and volumetric pipettes resist breakage. Each pipette has blue graduations, ring mark, and inscriptions. These plastic pipettes are ideal for chemistry and biology applications. They can be used as disposable or multi-use devices. When cleaning, it is recommended they be washed with mild alkaline detergent up to 60 °C. Exposure to temperatures above 60 °C may cause volume changes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty