Would you like to log in again?
Budget Tabletop Training IV Stand
Supplier: THE APPRENTICE CORPORATION
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AD604-STAND
470313-864EA 15.5 USD InStock
470313-864
Budget Tabletop Training IV Stand
Support Stands
A low cost stand for holding fluid supplies for simulators.
  • Holds blood or IV bags
  • Adjustable height
This two hook stand is a great accessory for any training program. Designed as a blood supply stand for a simulator, it can also be used to hold IV or blood bags for connection training. The stand has a 6" diameter base, and adjusts from 15" to 25" high. The base also has punch-outs, allowing users to attach suction cups (not included), or to attach the base to a surface using screws (not included) for additional stability when using at full extension.
Ordering information: Image may show items not included with purchase.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty