Would you like to log in again?
Glass Wool
Supplier: EISCO SCIENTIFIC LLC
GLW500
470313-974EA 57.2 USD InStock
470313-974
Glass Wool
Glass and Cotton Wool
For filtering out fine precipitates when filter papers are unsuitable.
  • Non-flammable
  • Provides thermal insulation
Non-flammable glass wool for use in general laboratory and industrial applications. Used for filtering out fine precipitates where filter papers are not suitable and the intertwined borosilicate glass fibers provide thermal insulation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty