Would you like to log in again?
Gooch Type Crucibles - Sintered
Supplier: EISCO SCIENTIFIC LLC
CH0465G CH0465H CH0465F
470313-998EA 17.9 USD InStock
470313-998 470313-996 470314-000
Gooch Type Crucibles - Sintered
Crucibles
Borosilicate glass with sintered disc of porosity G-3.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty