Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rock Forming Mineral Collection Series

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Ward's®
WARD470314-454 WARD470314-418 WARD470314-412 WARD470314-416 WARD470314-420 WARD470314-410 WARD470314-414
470314-416KT 29.6 USD InStock
470314-416 470314-418 470314-412 470314-414 470314-420 470314-454 470314-410
Rock Forming Mineral Collection Series
Specimens Mineral Specimens Mineral Collections
Excellet series for college mineralogy and petrology courses.

  • Hand cut by Ward's geologists
  • Specimens (72), 1×1 to 1×2", conveniently broken up into affordable collections

Allow your students to explore the rock-forming minerals typical of those found in standard textbooks. These sets are housed in compartmented collection boxes and come with information lists with localities.

Rock-forming Set 1: Includes 7 feldspar and 2 feldspathoid
Rock-forming Set 2: Includes 3 mica, 3 pyroxene, and 3 amphiboles
Rock-forming Set 3: Includes 2 olivine, 3 garnet, 2 epidote, 3 aluminosilicates, 1 tourmaline, and 1 topaz
Rock-forming Set 4: Includes 8 quartz, 3 hydrous silicate, and 1 chlorite
Rock-forming Set 5: Includes 2 serpentine, 1 talc, 1 zeolite, and 5 oxides and sulfides
Rock-forming Set 6: Includes 9 oxides and sulfides
Rock-forming Set 7: Includes 1 phosphate and 11 carbonates, sulfates, and halides

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty