Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ward's® Weigh Bucket
Supplier: MTC Bio
Ward's®
B6602-W-WARDS
470314-478CS 59.15 USD InStock
470314-478
Ward's® Weigh Bucket
Boats Weighing Boats
The weigh boat for liquids.
  • Easy-grip handle
  • 40 ml capacity
  • Convenient pour spout
  • Stackable
Ths polyethylene weigh bucket is a new type of weighing vessel that safely transports, contains and pours liquids, powders and small solids. The handle and spout make them easy to use, practically eliminating spillage and making controlled-rate pouring possible.
Caution: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty