Would you like to log in again?
Ward's® Pure Preserved Freshwater Perch
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Ward's®
WARD470314-678 WARD470314-676 WARD470314-616 WARD470314-550 WARD470314-762 WARD470314-654
470314-616PK 389.99 USD InStock
470314-616 470314-676 470314-654 470314-678 470314-550 470314-762
Ward's® Pure Preserved Freshwater Perch
Specimens Preserved Specimens Preserved Animal Materials
Ward's® PURE Preserved Specimens - the best dissection experience from start to finish.
  • Safest specimens
  • Low formalin odor
  • Best classroom experience
Ward's® PURE preserved specimens offer the best classroom experience because PURE specimens are safe. Students stay focused on learning and not on smells. Experience quality with true-to-life preservation, combined with artfully illustrated dissection guides, free with every purchase, to maximize value, lesson time and student engagement.
We guarantee you’ve never experienced dissection like this before. Go Pure.
Yellow perch, Perca flavescens are bony fishes of the class osteichthyes. Perch make a great dissection for middles chool, high school and college students alike. Introduce topics like vertebrate anatomy and bony fish anatmony. Ideal for studying external anatomical features, internal organs and organ systems. Plain and single-injected specimens are available. Single injected specimens are injected with red latex to highlight the arterial system.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty