Would you like to log in again?
LENS SEMICIRCULAR GLASS 90MM/45MM RADIUS
Supplier: SCIENCE INTERACTIVE GROUP
Lens Semicircular Glass 90Mm/45Mm Radius^ Semicircular glass lens, 90mm base, 45 mm radius. Transparent on one side and frosted on other side
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LENS SEMICIRCULAR GLASS 90MM/45MM RADIUS 470314-996 614-0600 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty