Would you like to log in again?
Ward's® Petrology Collection of Rocks and Minerals Series
Supplier: VWR
Ward's®
WARD470315-002 WARD470315-000 WARD470315-004
470315-004KT 76.8 USD InStock
470315-004 470315-002 470315-000
Ward's® Petrology Collection of Rocks and Minerals Series
Specimens Rock Specimens Rock Collections
Advanced study series of almost 100 specimens.
  • Hand cut by Ward's geologists
  • Includes 96 specimens, 1×1 to 1×2", conveniently broken up into affordable collections
Well-defined samples of rocks and rock-forming minerals that are idea for study. Students will be able to identify important textures and structures in the carefully selected specimens. Each set comes in a house in a compartmented collection box with an identification list including localities.
Igneous Collection: Includes 18 igneous rocks and 18 igneous minerals.
Sedimentary Collection: Includes 22 sedimentary rocks and 14 sedimentary minerals.
Metamorphic Collection: Includes 13 metamorphic rocks and 11 metamorphic minerals.
Complete Petrology Series: Buy all three sets and save.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty