Would you like to log in again?
Wards® STEM Investigations: Create Your Own Power
Supplier: ALDON CORP SE
Wards®
IS3450
470315-144KT 445 USD InStock
470315-144
Wards® STEM Investigations: Create Your Own Power
Educational Materials Chemistry Educational Materials Electrochemistry Learning Activities
Set up an electrochemical cell using a variety of materials.
  • Includes two different types of electrochemical cells: salt bridge and semi-permeable membrane
The voltaic cell is an electrochemical cell that produces electrical energy through a chemical reaction, specifically an oxidation-reduction reaction. In this activity, students will set up electrochemical cells, with each group using a different combination of materials for the anode and the cathode of the cell. Students will also compare two different styles of cell, one using a salt bridge and one using a semi-permeable membrane. After determining the voltage output of their cells and comparing their values to the rest of the class, students will be challenged to design an electrochemical cell that can run a small motor. Kit contains enough materials for 15 groups. Teacher’s Manual and Student Study Guide copymasters are included.
Delivery information: Includes 1 L Copper chloride 1.0 M; 1 L Magnesium chloride 1.0 M; 1 L Iron chloride 1.0 M; 1 L Tin (II) chloride 1.0 M; 1 L Zinc chloride 1.0 M; 1 L Aluminum chloride 1.0 M; 1 L Potassium chloride 1.0 M; 125 pieces Copper metal foil strips; 125 pieces Tin foil strips; 125 pieces Iron metal strips; 125 pieces Aluminum metal strips; 125 pieces Zinc strips; 125 pieces Magnesium metal ribbon strips; 60 cups, 5 oz; 15' Dialysis tubing;15 Red connecting wires with alligator clips; 15 Black connecting wires with aligator clips; 20 pieces Filter paper; 60 Pipettes; 15 3 V Motors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty