Would you like to log in again?
Dissecting Forceps
Supplier: DR INSTRUMENTS INC. SE
20L 15C
470315-192BX 185 USD InStock
470315-192 470315-182
Dissecting Forceps
Forceps
Quality fine point forceps.
  • Straight or bent tip
  • Stainless steel construction
  • Textured grip
Dissecting forceps feature straight fine points. Made with high quality stainless steel. Fine points are great for delicate dissection purposes and can be used under the microscope.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty