Would you like to log in again?
Tissue Forceps
Supplier: DR INSTRUMENTS INC. SE
19-T
470315-188BX 222 USD InStock
470315-188
Tissue Forceps
Forceps
Hold slippery tissues easily.
  • Stainless steel construction
  • Made with 1×2 teeth
These large forceps are perfect for holding difficult tissue. The longer size gives you room to move and plenty of leverage. The tip uses three teeth to hold tissues securely. As these features may damage some tissues, these are usually used during initial dissection, and not with organs.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty