Would you like to log in again?
Featherweight Forceps Set
Supplier: DR INSTRUMENTS INC. SE
DRENTF-II
470315-238BX 163 USD InStock
470315-238
Featherweight Forceps Set
Forceps
Use a soft touch for delicate specimens.
  • Light closure for delicate specimens
Featherweight entomology forceps set with one narrow tip and one wide tip forceps. Made out of flexible stainless steel (Spring Stainless Steel), these forceps can be bent virtually in any shape and form. These forceps are ideal for handling larvae or eggs without crushing them and specimens stored in vials.
Delivery information: Set includes broad and fine points and plastic storage case.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty