Would you like to log in again?
Microdissection Forceps Set
Supplier: DR INSTRUMENTS INC. SE
MDFKIT-1
470315-242BX 113 USD InStock
470315-242
Microdissection Forceps Set
Forceps
Precision cut points.
  • Includes five varied tip 4.5 in forceps
  • Extra fine points
  • Storage case
Micro-dissection forceps set contains widely used five pairs of forceps. Each pair of forceps have highly precise and extra fine tips (points) to perform micro-dissection and or any fine work under the microscope. This kit includes two curved and three straight micro-dissection forceps
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty