Would you like to log in again?
Ward's® Essentials Stereomicroscope
Supplier: VWR
Ward's®
SE306R-P-LED
470315-512EA 160.35 USD InStock
470315-512
Ward's® Essentials Stereomicroscope
Microscopes
Versatile and durable, this stereomicroscope is ideal for the high school classroom.
  • 20× and 40× magnifications
  • Interchangeable stage plates: translucent and black/white
  • Adjustable interpupillary distance
  • One year limited warranty
  • Rechargeable LED illumination
Features include 45° inclined viewing head, post mount that adjusts the head up or down to accommodate larger objects on stage and easy-adjust collar for tension control.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty