Would you like to log in again?
Fracture Collection
Supplier: VWR
Ward's®
WARD470315-516
470315-516KT 21.99 USD InStock
470315-516
Fracture Collection
Specimens Mineral Specimens Mineral Collections
Demonstrates irregular breakage of minerals.
Each specimen illustrates a type of mineral breakage that does not leave a smooth cleavage surface. The set is housed in a compartmented collection box, and an identification list is included.
Contents: 6 numbered samples, 1×1"
Even Fracture - Halite
Uneven Fracture - Agate
Conchoidal Fracture - Obsidian
Subconchoidal Fracture - Rose Quartz
Splintery Fracture - Satin Spar Gypsum
Hackly Fracture - Copper
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty