Would you like to log in again?
ADC® EKG Ruler
Supplier: AMERICAN DIAGNOSTIC CORP CP
ADC®
394
470315-770CS 10901.25 USD InStock
470315-770
ADC® EKG Ruler
Rulers
A powerful addition for EKG measurement.
  • Seconds scale
  • Offers both one and two cycle scales
  • Small enough to fit in a medical coat pocket
  • Complete with protective sleeve
Quickly and easily determine rates from an EKG with this convenient tool. The seconds scale is readily used for PR, QRS, cycle, length, coupling, escape and parasystalic intervals. Easily protected and stored in a clear sleeve, don’t need to worry about spills or splashes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty