Would you like to log in again?
Baseline® X-ray Goniometers
Supplier: AMERICAN 3B SCIENTIFIC SE
3009526 3009525 3009527
470316-696EA 64.99 USD InStock
470316-696 470316-698 470316-694
Baseline® X-ray Goniometers
Goniometers
Easily measure angles on negatives.
  • Anodized finish
  • 180° measurement
  • Radiopaque markings
The Baseline X-ray goniometer can document angles on X-ray negatives. 180° head has two opposing scales marked in 21/2° increments. Black anodized aluminum with easy-to-read white radiopaque markings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty