Would you like to log in again?
DC Power Supply
Supplier: AMERICAN 3B SCIENTIFIC SE
1012857
470316-706EA 586.59 USD InStock
470316-706
DC Power Supply
Power Supplies
High-quality switched-mode power supply in space-saving housing with intelligent control of fan speed to ensure safe and quiet operation.
  • DC output 1 – 32 V, 0 - 20 A (0 – 5 A output with 4-mm safety sockets on front)
  • 640 W max. output power
  • 3-digit, 15 mm, green LED display
Simple, precise and fast adjustment of voltage and current levels with dual-function rotary knobs for coarse and fine adjustment. Adjustable current limiting in open circuit. Three user-definable stored configurations for voltage and current limiting make it easy to recall frequently used settings. Full remote control of voltage and current plus output which can be turned on and off.
Dimensions: 200×90×255 mm
Weight: 2.6 kg
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty