Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Experimental Safety Leads Set

Supplier: AMERICAN 3B SCIENTIFIC SE
1002841
470316-710EA 130 USD InStock
470316-710
Experimental Safety Leads Set
Power Supplies
For electrical circuits with low voltages and high currents; copper wire encased in highly flexible PVC; equipped at either end with a laminated plug and a fully-insulated axial jack for connecting additional patch cords.

  • Wire Cross-Section: 2.5 mm2
  • Continuous Current: Maximum. 32 A
  • Plug and Jack: 4 mm (Nickel-Plated)

Set of 15 patch cords, 75 cm long, 5 of each color in red, black, and blue.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty