Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Digital Orbital Shaker

Supplier: Heathrow Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

120460
470316-816EA 1275.65 USD InStock
470316-816
Digital Orbital Shaker
Shakers and Mixers Orbital Shakers
Versatile shaker for a vast range of protocols. Create own matrix for vessels of all shapes and sizes with Hi-Lo platform and bungee cords.

  • Last run memory, enables reproducibe results
  • Has a removable non-skid silicone mat to help hold vessels securely in place
  • ABS body resist most commonly used chemicals and is easy to clean
  • Quiet operation
  • Back-lit Display
  • Two control knob
  • Spill-resistant tray
  • Applications include incubators, cold rooms, cell cultures, immunoassays, and protein studies

Soft-start feature slowly increases to desired speed to avoid splashing. Compact design saves bench top space and fits most incubators.

No-stop electric motor feedback enables the unit to instantly adjust to weight load changes and unbalanced loads for truly reproducible results. Shut-off feature for higher speeds reduces spill occurrences from full vessels.

Time range: 0 to 74 hours and 59 minutes, 1 second increments or continuous.

Max. Relative Humidity: Non-condensing, 80% for temperatures up to 31 °C decreasing linearly to 50% R.H. at 40 °C.

Certifications: CE, UKCA, Intertek (US, CAN), ROHS3, and WEEE.

Ordering information: Five-year warranty.

Delivery information: Includes Shaker, skid-free rubber platform mat, 8 bungee cord set, Universal Hi-Lo platform with soft rubber edge bumpers, 1 low voltage, double insulated power adapter (110 to 240 VAC, 50/60 Hz) with 4 interchangeable plugs.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty