Would you like to log in again?
Kidder Gear System
Supplier: KIDDER MFG. CO. LTD
Ward's®
83-54-GSCP
470316-954EA 256.55 USD InStock
470316-954
Kidder Gear System
Educational Materials Physics Educational Materials Mechanics Learning Activities
Gears are so much a part of everyday life that it would be easy to overlook their impact.
  • There are enough materials in this classpack to make 12 kits
  • Requires minimal drilling, cutting and gluing
This NEW Kidder designed kit is the perfect project for your students to investigate ways in which gears modify the speed and direction of, and the force exerted on moving objects. After building and assembling this kit, your students will have an understanding of the basic principles and functions of gear systems. Watch one movement propelled by a DC motor and worm gear turn 11 different gears to demonstrate differentials and ratios in a clear and concise fashion. Put your students in groups of two and get building! There are thorough instructions provided with this kit that also mention ties to curriculum and every day life. A great project to work on in your MakerSpace, lab or library!
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty