Would you like to log in again?
Visual Scientifics Photogate System, Eisco Scientific
Supplier: Eisco Scientific
PTPGS
470317-428EA 353.95 USD InStock
470317-428
Visual Scientifics Photogate System, Eisco Scientific
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
A complete photogate system for use with Visual Scientifics systems - or any lab experiment requiring photogates.
  • Instruction manual included
  • Platform neutral - works with any physics apparatus
This photogate system is an accessory for use with Visual Scientifics experiment kits, but it may be used for many other applications as well. This tool has a built in stopwatch, and features four function modes: gate, pulse, pendulum and frequency.
Ordering information: Please note that the Visual Scientifics experiment kits and components (including the magnetic base) are not included with the photogate system.
Delivery information: The system includes photogate timer, timer power cord, 2 photogates, 2 photogate cables, 2 support poles, 2 magnetic pole bases, and 2 sliding pole supports.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty