Would you like to log in again?
Bone Clones® Australopithecus boisei Cranium, Female KNM ER 732
Supplier: BONE CLONES, INC.
BH-026
470317-796EA 270 USD InStock
470317-796
Bone Clones® Australopithecus boisei Cranium, Female KNM ER 732
Models Human Biology Models
1.7 MYA. Female. The Australopithecus boisei skull KNM ER 732 was discovered in 1970 at Koobi Fora, Kenya by R. Leakey and H. Mutua and described in Nature in 1971.
  • Each Bone Clones® cast is hand-made from start to finish
  • Rigorous standards are applied when selecting original specimens to ensure the best representation
  • Proprietary, custom blend of polyurethane developed to capture and replicate minute detail
A comparative analysis of KNM ER 732 and KNM ER 406 revealed a host of similarities with the only differences (e.g., smaller size, lack of sagittal and nuchal crests in KNM ER 732) being typical of the sexual dimorphism found in great ape skulls. Consequently, KNM ER 732 is considered to be a female and KNM ER 406 a male of the species, Australopithecus boisei. Using the sexual dimorphism found in this pair as a guide, researchers suggested that A. robustus and A. africanus, which had been hypothesized by some to be male and female specimens of the same species, were, in fact, far more different than one would expect from simple sexual dimorphism. They proposed instead that A. robustus and A. africanus were separate species.
The Bone Clones® Hominid line is composed of discoveries from anatomically modern humans, archaic humans, early Homo, early hominins, and other hominids. The majority of the casts in this line have been recreated by a team of anatomical sculptors. Some are reconstructions made by anthropology professionals using fragmentary elements from original discoveries and extrapolating the missing parts from those.
Accessories information: Accessories not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty