Would you like to log in again?
Cardionics® SAM Basic Auscultation Manikin
Supplier: AMERICAN 3B SCIENTIFIC SE
1022474 1020097 3012368 3012373 1020099
470318-062EA 1800.09 USD InStock
470318-062 470317-928 470318-054 470317-924 470318-058
Cardionics® SAM Basic Auscultation Manikin
Models Human Biology Models
Easily learn auscultation.
  • Includes COVID sounds
  • Lightweight and portable
  • Extensive cardionics proprietary Heart, Lung, Bowel and Bruit sounds library
  • Students can listen at anatomically correct auscultation sites
Auscultation is an essential clinical skill needed to assess and monitor patient's conditions. With Cardionics SAM Basic Student Auscultation Manikin, teaching and learning this skill becomes even more versatile and economic. SAM Basic trainer can be used with the SimScope™ stethoscope (included). All sounds and videos are recorded from adult live patients for students to experience a life-like simulation.
SAM Basic Student Auscultation Manikin used in teaching and learning heart, lung and bowel sounds. The SimScope™ stethoscope is used to simulate adult Heart, Lung and bowel sounds, utilizing a large sound library incorporated into the SimScope stethoscope.
Accessories information: See additional information for feature details.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty